Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethanol 95.0+%, TCI America™
SDP

Catalog No. B0590100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0590100G 100 g
B059025G 25 g
B0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0590100G Supplier TCI America Supplier No. B0590100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-51-2
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00002827
InChI Key LDLCZOVUSADOIV-UHFFFAOYSA-N
Synonym ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
PubChem CID 10898
IUPAC Name 2-bromoethanol
SMILES C(CBr)O

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 150°C
Quantity 100 g
UN Number 2922
Formula Weight 124.97
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethanol
EINECSNumber (2)-2002
RTECSNumber KJ8225000
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.