Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Bromoethoxy)-tert-butyldimethylsilane

Catalog No. 42842610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-842-610G 10 g
42-842-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-842-610G Supplier Sigma Aldrich Supplier No. 42842610G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2-Bromoethoxy)-tert-butyldimethylsilane is a silane derivative.(2-Bromoethoxy)-tert-butyldimethylsilane was used in the synthesis of 4-(3-hydroxypropyl)-4'-methyl-2,2'-bipyridine.It may be used as a reagent for the selective N-alkylation of 5-piperazin-1-yl-1H-indole and (1H-indol-2-yl)-piperazin-1-yl-methanone and also in the synthesis of 2-[3-[(3,4,5-trimethoxyphenyl)thio]-1H-indol-5-yloxy]ethanol.

Specifications

CAS 86864-60-0
Density 1.115 g/mL (at 25°C (literature))
Boiling Point 70°C to 75°C (2.5 mmHg)
Molecular Formula C8H19BrOSi
Refractive Index n20/D 1.444 (literature)
Linear Formula BrCH2CH2OSi(CH3)2 C(CH3)3
MDL Number MFCD00209550
Quantity 10 g
Molecular Weight (g/mol) 239.23
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.