Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethyl Chloroformate 97%, Thermo Scientific™

Catalog No. AC209830250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC209830250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC209830250 Supplier Thermo Scientific Chemicals Supplier No. 209830250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 4801-27-8
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H4BrClO2
Refractive Index 1.4770 to 1.4790 (20°C, 589nm)
Linear Formula ClCO2CH2CH2Br
Quantity 25g
Synonym 2-bromoethyl chloroformate, 2-bromoethylchloroformate, 2-bromoethyl chlorocarbonate, carbonochloridic acid, 2-bromoethyl ester, 2-bromoethoxy carbonyl chloride, chloroformic acid 2-bromoethyl ester, 2-bromoethyl chloridocarbonate, .beta.-bromoethyl chloroformate, ethanol, 2-bromo-, chloroformate
InChI Key ZLCKDYMZBZNBMJ-UHFFFAOYSA-N
SMILES C(CBr)OC(=O)Cl
IUPAC Name 2-bromoethyl carbonochloridate
Molecular Weight (g/mol) 187.417
PubChem CID 78528
Formula Weight 187.42
Percent Purity ≥96.0%
Chemical Name or Material 2-Bromoethyl chloroformate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 225-361-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.