Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethyl Phenyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. B20855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20855G 5 g
B208525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20855G Supplier TCI America Supplier No. B20855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4837-01-8
Molecular Formula C8H9BrS
Molecular Weight (g/mol) 217.12
MDL Number MFCD00040826
InChI Key UEFBOQYLXLEJSM-UHFFFAOYSA-N
Synonym 2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, acmc-1aoyt, 2-phenylthioethyl bromide
PubChem CID 78553
IUPAC Name [(2-bromoethyl)sulfanyl]benzene
SMILES BrCCSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 217.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl Phenyl Sulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.