Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromoethylamine hydrobromide

Catalog No. B65705100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B65705100G 100 g
B6570525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B65705100G Supplier Sigma Aldrich Supplier No. B65705100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromoethylamine hydrobromide

Specifications

CAS 2576-47-8
Melting Point 170°C to 175°C (lit.)
Molecular Formula C2H6BrN -+ HBr
Linear Formula BrCH2CH2NH2 -+ HBr
MDL Number MFCD00012886
Quantity 100 g
Synonym 2-Aminoethyl bromide hydrobromide
Molecular Weight (g/mol) 204.89
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.