Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethylamine hydrobromide, 99%

Catalog No. AC106930010
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106930010 1 kg Plastic bottle
AC106930250 25 g Plastic bottle
AC106931000 100 g Plastic bottle
AC106932500 250 g Plastic bottle
AC106935000 500 g Plastic bottle
AC106930025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AC106930010 Supplier Thermo Scientific Chemicals Supplier No. 106930010
  • $1,032.00
    $841.65 / Each of 1

    Save $190.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2576-47-8 , 107-09-5
Molecular Formula C2H6BrN·HBr
Molecular Weight (g/mol) 204.9
MDL Number MFCD00012886
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula BrCH2CH2NH2·HBr
Quantity 1 kg
Beilstein 04,134
Solubility Information Solubility in water: >500g/L (20°C)
Formula Weight 204.9
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromoethylamine hydrobromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 219-924-2
RTECSNumber KQ8225000
TSCA TSCA
Recommended Storage this product has a strong avidity for water

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.