Learn More
Supelco Inc 2-Bromoheptane, 1974-04-5, MFCD00000164, 25 G
Supplier:  Supelco Inc 29238925G
Encompass
Linear Formula: CH3CH2CH2CH2CH2CHBrCH3, Molecular Weight: 179.10, Synonym: 1-Methylhexyl bromide, Technical Grade, BP: 64-66 °C/21 mmHg(lit.). 2-Bromoheptane has been used in the preparation of: racemic alcohol, 2-methylheptanol, glutathione (GSH) derivatives, GSH analog that contains tetrazole isostere (Tet-sHep).
Catalog No. NC0877547