Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromohexane, MP Biomedicals

Catalog No. ICN205336 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205336 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205336 Supplier MP Biomedicals, Inc Supplier No. 0520533605

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3377-86-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
InChI Key NEBYCXAKZCQWAW-UHFFFAOYSA-N
Synonym hexane, 2-bromo, 2-hexyl bromide, dl, 2-bromo-hexane, acmc-209t5g, nebycxakzcqwaw-uhfffaoysa
PubChem CID 18805
IUPAC Name 2-bromohexane
SMILES CCCCC(C)Br

Specifications

Quantity 5 g
Formula Weight 165.08
Chemical Name or Material 2-Bromohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.