Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoisobutyramide 98.0+%, TCI America™
SDP

Catalog No. B33805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33805G 5 g
B338025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33805G Supplier TCI America Supplier No. B33805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7462-74-0
Molecular Formula C4H8BrNO
Molecular Weight (g/mol) 166.018
MDL Number MFCD08234837
InChI Key WTRIMJTZOOLIFZ-UHFFFAOYSA-N
Synonym 2-Bromo-2-methylpropionamide
PubChem CID 101235
IUPAC Name 2-bromo-2-methylpropanamide
SMILES CC(C)(C(=O)N)Br

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.02
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoisobutyramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.