Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Bromoisonicotinic acid | 66572-56-3 | 202.01 | 25 G
SDP

Catalog No. 5000448286
Encompass_Preferred

2-Bromoisonicotinic acid is a non-coding amino acid-based herbicide. It is intended for research use only.

  • Purity: 99.86%
  • CAS number: 66572-56-3
  • Molecular weight: 202.01
  • Formula: C6H4BrNO2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-48286 Supplier Medchemexpress LLC Supplier No. 25GHYW002805
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Bromoisonicotinic acid is a non-coding amino acid-based herbicide. It is intended for research use only.

  • Purity: 99.86%
  • CAS number: 66572-56-3
  • Molecular weight: 202.01
  • Formula: C6H4BrNO2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.