Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromomethyl-1,4-benzodioxane 98.0+%, TCI America™
SDP

Catalog No. B23491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23491G Supplier TCI America Supplier No. B23491G

Chemical Identifiers

CAS 2164-34-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00044831
InChI Key QYLFKNVZIFTCIY-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,4-benzodioxane, 2-bromomethyl-2,3-dihydro-1,4-benzodioxine, 2-bromomethyl-2,3-dihydrobenzo b 1,4 dioxine, 2-bromomethyl-2,3-dihydro-benzo 1,4 dioxine, 2-bromomethyl-2,3-dihydrobenzo 1,4 dioxine, 2-bromomethyl-2,3-dihydro-1,4-benzodioxin, 3-bromomethyl-2,3-dihydro-1,4-benzodioxine, acmc-1cuiw, 2-bromomethylbenzo-1,4-dioxan
PubChem CID 98333
IUPAC Name 3-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
SMILES C1C(OC2=CC=CC=C2O1)CBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromomethyl-1,4-benzodioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.