Learn More
Medchemexpress LLC 2-Bromonaphthalene-d7 | 1359833-48-9 | 214.11 | 50 MG
SDP
Catalog No. 5000445809
Encompass_Preferred
2-Bromonaphthalene-d7 is a deuterium-labeled version of 2-Bromonaphthalene, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is being studied for its potential to influence pharmacokinetic and metabolic profiles of drugs. The compound appears as a solid with an off-white to light yellow color.
  • Deuterium-labeled compound
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, and LC-MS techniques
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
  • Appears as an off-white to light yellow solid
Catalog No. 50-004-45809 Supplier Medchemexpress LLC Supplier No. HYW007537S50MG
2-Bromonaphthalene-d7 is a deuterium-labeled version of 2-Bromonaphthalene, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is being studied for its potential to influence pharmacokinetic and metabolic profiles of drugs. The compound appears as a solid with an off-white to light yellow color.
  • Deuterium-labeled compound
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, and LC-MS techniques
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
  • Appears as an off-white to light yellow solid