Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromonicotinic acid, 97%

Catalog No. AAH3023903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3023903 1 g
AAH3023906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3023903 Supplier Thermo Scientific Chemicals Supplier No. H3023903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 35905-85-2 | C6H4BrNO2 | 202.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35905-85-2
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646068
InChI Key MMWNKXIFVYQOTK-UHFFFAOYSA-N
Synonym 2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f
PubChem CID 233262
IUPAC Name 2-bromopyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Br)C(=O)O

Specifications

Melting Point ∼254°C
Quantity 1 g
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 2-Bromonicotinic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.