Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
- Sigma-aldrich does not collect analytical data for this product
- Buyer assumes responsibility to confirm product identity and/or purity
- All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise.product of chembridge corp
Specifications
Specifications
| CAS | 1074-16-4 |
| Density | 1.483 g/mL (at 25°C (literature)) |
| Boiling Point | 97°C (0.7 mmHg) |
| Molecular Formula | C8H9BrO |
| Refractive Index | n20/D 1.577 (literature) |
| Linear Formula | BrC6H4CH2CH2OH |
| Quantity | 1 g |
| MDL Number | MFCD00093566 |
| Molecular Weight (g/mol) | 201.06 |
| Percent Purity | 99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?