Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B47775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47775G Supplier TCI America Supplier No. B47775G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129112-25-0
Molecular Formula C7H4BrF3O3S
MDL Number MFCD18379777
Synonym Trifluoromethanesulfonic Acid 2-Bromophenyl Ester, 2-Bromophenyl Triflate

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 g
Formula Weight 305.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromophenyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.