Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromophenyl)urea 98.0+%, TCI America™
SDP

Catalog No. B28335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28335G Supplier TCI America Supplier No. B28335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13114-90-4
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
InChI Key QXEDXIJDCOADGG-UHFFFAOYSA-N
PubChem CID 139386
IUPAC Name (2-bromophenyl)urea
SMILES C1=CC=C(C(=C1)NC(=O)N)Br

Specifications

Melting Point 202°C
Color Red-White
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (2-Bromophenyl)urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.