Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenylacetonitrile, 97%

Catalog No. AAA1754814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1754814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1754814 Supplier Thermo Scientific Chemicals Supplier No. A1754814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 19472-74-3 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19472-74-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001896
InChI Key BVCOJESIQPNOIF-UHFFFAOYSA-N
Synonym 2-bromophenylacetonitrile, 2-bromobenzyl cyanide, 2-2-bromophenyl acetonitrile, o-bromobenzyl cyanide, 2-bromophenyl acetonitrile, o-bromophenylacetonitrile, benzeneacetonitrile, bromo, 2-2-bromophenyl ethanenitrile, benzeneacetonitrile, 2-bromo, 2-bromo-phenyl-acetonitrile
PubChem CID 29625
IUPAC Name 2-(2-bromophenyl)acetonitrile
SMILES BrC1=CC=CC=C1CC#N

Specifications

Melting Point 19°C
Density 1.51
Boiling Point 252°C
Flash Point 113°C (235°F)
Odor Pungent
Refractive Index 1.5696
Quantity 25 g
UN Number UN1694
Beilstein 2043684
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 2-Bromophenylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BROMOBENZYL CYANIDES, LIQUID
EINECSNumber 243-091-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.