Learn More
Strem, An Ascensus Company CAS# 62336-24-7. 1g. 2-Bromophenyldiphenylphosphine, 98%. MFCD00045014
SDP
Catalog No. 5090109768
Encompass
CAS# 62336-24-7. 1g. 2-Bromophenyldiphenylphosphine, 98%. MFCD00045014. Molecular Weight: 341.18. Molecular Formula: C18H14BrP. Color/form: white pwdr. Strem# 15-0125. http://www.strem.com/catalog/v/15-0125/
Catalog No. 50-901-09768 Supplier Strem, An Ascensus Company Supplier No. 1501251G
CAS# 62336-24-7. 1g. 2-Bromophenyldiphenylphosphine, 98%. MFCD00045014. Molecular Weight: 341.18. Molecular Formula: C18H14BrP. Color/form: white pwdr. Strem# 15-0125. http://www.strem.com/catalog/v/15-0125/