Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromopropane

Catalog No. 501861312
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-1312 100 g
50-186-1313 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-186-1312 Supplier Sigma Aldrich Supplier No. B78114100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromopropane
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-26-3
Melting Point -89°C (lit.)
Density 1.31 g/mL (at 25°C (literature))
Boiling Point 59°C (lit.)
Molecular Formula C3H7Br
Refractive Index n20/D 1.425 (literature)
Linear Formula (CH3)2 CHBr
MDL Number MFCD00000147
Quantity 100 g
Synonym Isopropyl bromide
Molecular Weight (g/mol) 122.99
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.