Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopropionic acid, 98%

Catalog No. AAA1756930
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1756930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1756930 Supplier Thermo Scientific Chemicals Supplier No. A1756930
  • $83.60
    $71.65 / Each of 1

    Save $11.95

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 598-72-1 | C3H5BrO2 | 152.98 g/mol

It is employed as pharmaceutical and agrochemical intermediate. It is used as an alkylation agent for mercaptans and other sulfur containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as pharmaceutical and agrochemical intermediate. It is used as an alkylation agent for mercaptans and other sulfur containing compounds.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 598-72-1
Melting Point 25°C
Density 1.7
Boiling Point 202°C to 203°C
Flash Point 100°C (212°F)
Molecular Formula C3H5BrO2
Refractive Index 1.476
Quantity 250 g
MDL Number MFCD00004211,MFCD00145205
UN Number UN3261
Beilstein 605451
Synonym 2-bromopropionic acid, propanoic acid, 2-bromo, dl-2-bromopropionic, propionic acid, 2-bromo, alpha-bromopropionic acid, dl-2-bromopropionic acid, 2-bromo-propionic acid, .alpha.-bromopropionic acid, propanoic acid, bromo, propanoicacid, 2-bromo
Solubility Information Soluble in water.
InChI Key MONMFXREYOKQTI-UHFFFAOYNA-N
SMILES CC(Br)C(O)=O
IUPAC Name 2-bromopropanoic acid
Molecular Weight (g/mol) 152.98
PubChem CID 11729
Formula Weight 152.98
Percent Purity 98%
Chemical Name or Material 2-Bromopropionic acid
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-947-6
RTECSNumber UA2451715
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.