Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromopropionyl bromide

Catalog No. 249785500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-978-5500G 500 g
24-978-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-978-5500G Supplier Sigma Aldrich Supplier No. 249785500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromopropionyl bromide has been used in the synthesis of:
  • Cellulose macroinitiator via direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride
  • 3-(2-bromopropionyl)-2-oxazolidone derivatives
  • Bromoester terminated poly(3-hexylthiophene), used as macroinitiator for atom transfer radical polymerization of acrylates

Specifications

CAS 563-76-8
Density 2.061 g/mL (at 25°C (literature))
Boiling Point 48°C to 50°C (10 mmHg)
Molecular Formula C3H4Br2O
Refractive Index n20/D 1.518 (literature)
Linear Formula CH3CHBrCOBr
MDL Number MFCD00000142
Quantity 500 g
Molecular Weight (g/mol) 215.87
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.