Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromopyridine

Catalog No. B80100500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B80100500G 500 g
B80100100G 100 g
B8010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B80100500G Supplier Sigma Aldrich Supplier No. B80100500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromopyridine can be used as:
  • A building block in the formation of cn bond by various cross coupling reactions
  • A reactant in negishi cross-coupling reaction with aryl halides catalyzed by palladium
  • A reactant in the synthesis of 2α-pyridyldifluoroacetate with ethyl bromodifluoroacetate in presence of copper as a catalyst

Specifications

CAS 109-04-6
Density 1.657 g/mL (at 25°C (literature))
Boiling Point 192°C to 194°C (lit.)
Molecular Formula C5H4BrN
Linear Formula C5H4BrN
Refractive Index n20/D 1.572 (literature)
MDL Number MFCD00006219
Quantity 500 g
Molecular Weight (g/mol) 158
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.