Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B51701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51701G 1 g
B5170-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51701G Supplier TCI America Supplier No. B51701G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Bromopyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 452972-08-6
Melting Point 132°C
Color White-Yellow
Molecular Formula C5H5BBrNO2
Quantity 1 g
MDL Number MFCD04113805
Synonym 2-bromopyridine-3-boronic acid, 2-bromopyridin-3-yl boronic acid, 2-bromo-3-pyridineboronic acid, 2-bromopyridine-3-ylboronic acid, 2-bromo-3-pyridylboronic acid, 2-bromopyridin-3-yl-3-boronic acid, 2-bromo-3-pyridyl boronic acid, 2-bromopyridine-3-boronicacid, pubchem5080, acmc-209k1y
InChI Key ODWRFKJNUJLAHM-UHFFFAOYSA-N
SMILES B(C1=C(N=CC=C1)Br)(O)O
IUPAC Name (2-bromopyridin-3-yl)boronic acid
Molecular Weight (g/mol) 201.814
PubChem CID 12067058
Formula Weight 201.81
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.