Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromostyrene, Stabilized 98%, Thermo Scientific™

Catalog No. AC221990010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221990010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC221990010 Supplier Thermo Scientific Chemicals Supplier No. 221990010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-88-5
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
InChI Key SSZOCHFYWWVSAI-UHFFFAOYSA-N
Synonym 2-bromostyrene, 1-bromo-2-vinylbenzene, o-bromostyrene, benzene, 1-bromo-2-ethenyl, unii-9a96gp6z5d, styrene, o-bromo, 2-bromo-styrene, o-bromovinylbenzene, pubchem23975, 2-bromo-1-vinylbenzene
PubChem CID 16264
IUPAC Name 1-bromo-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Br

Specifications

Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.5910 to 1.5930 (20°C, 589nm)
Linear Formula H2C=CHC6H4Br
Quantity 1 g
Beilstein 05, III, 1176
Stabilizer 0.005% 4 tert.-butylcatechol
Formula Weight 183.05
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Bromostyrene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.