Learn More
Medchemexpress LLC 2-Bromothiazole | 3034-53-5 | 164.02 | 25 G
SDP
Catalog No. 5000450047
Encompass_Preferred
2-Bromothiazole is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research purposes only. It is also utilized as an Aryl halide for N-arylation of 5- and 7-azaindoles, and copper-catalyzed cyanation provides 2-cyanothiazole.
  • Used as a biochemical reagent
  • Applicable as a biological material or organic compound in life science research
  • Can be used as an Aryl halide
Catalog No. 50-004-50047 Supplier Medchemexpress LLC Supplier No. 25GHYW001223
2-Bromothiazole is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research purposes only. It is also utilized as an Aryl halide for N-arylation of 5- and 7-azaindoles, and copper-catalyzed cyanation provides 2-cyanothiazole.
  • Used as a biochemical reagent
  • Applicable as a biological material or organic compound in life science research
  • Can be used as an Aryl halide