Learn More
Medchemexpress LLC 2-Bromothiazole | 3034-53-5 | 164.03 | 50 G
SDP
Catalog No. 5000448857
Encompass_Preferred
2-Bromothiazole is a biochemical reagent suitable for life science research. It serves as an organic compound or biological material and is intended for research purposes only. This aryl halide can be N-arylated with 5- and 7-azaindoles, and its copper-catalyzed cyanation yields 2-cyanothiazole.
  • Used as a biological material or organic compound for life science research
  • Intended for research purposes only
  • Functions as an aryl halide for N-arylation with 5- and 7-azaindoles
  • Can undergo copper-catalyzed cyanation to produce 2-cyanothiazole
Catalog No. 50-004-48857 Supplier Medchemexpress LLC Supplier No. HYW00122350G
2-Bromothiazole is a biochemical reagent suitable for life science research. It serves as an organic compound or biological material and is intended for research purposes only. This aryl halide can be N-arylated with 5- and 7-azaindoles, and its copper-catalyzed cyanation yields 2-cyanothiazole.
  • Used as a biological material or organic compound for life science research
  • Intended for research purposes only
  • Functions as an aryl halide for N-arylation with 5- and 7-azaindoles
  • Can undergo copper-catalyzed cyanation to produce 2-cyanothiazole