Learn More
Medchemexpress LLC 2-Bromothiazole-5-carboxylic acid | 54045-76-0 | 99.8% | 208.04 | 5 G
SDP
Catalog No. 5000404470
Encompass_Preferred
2-Bromothiazole-5-carboxylic acid is a starting compound used in the synthesis of active compounds. It can be used in the research of colon cancer, lung cancer, and liver cancer.
  • Used in the synthesis of NAMPT/HDAC inhibitors
  • Suitable for research into various cancers
  • Provided as a solid with light yellow to light brown appearance
Catalog No. 50-004-04470 Supplier Medchemexpress LLC Supplier No. HY332875G
2-Bromothiazole-5-carboxylic acid is a starting compound used in the synthesis of active compounds. It can be used in the research of colon cancer, lung cancer, and liver cancer.
  • Used in the synthesis of NAMPT/HDAC inhibitors
  • Suitable for research into various cancers
  • Provided as a solid with light yellow to light brown appearance