Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromothiophene

Catalog No. 12416850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-416-850G 50 g
12-416-8250G 250 g
12-416-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-416-850G Supplier Sigma Aldrich Supplier No. 12416850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromothiophene undergoes metalation-alkylation reaction with various electrophiles to form various 5-alkylated 2-bromo products
  • It undergoes coupling with 4-bromo allyl phenyl ether to form allyl phenyl thiophene ether, which is a novel potential dielectric material for organic thin film transistors.2-bromothiophene has been used in electrochemical reduction of a number of mono- and dihalothiophenes at carbon cathodes in dimethylformamide containing tetramethylammonium perchlorate by cyclic voltammetry and controlled-potential electrolysis

Specifications

CAS 1003-09-4
Density 1.684 g/mL (at 25°C (literature))
Boiling Point 149°C to 151°C (lit.)
Molecular Formula C4H3BrS
Refractive Index n20/D 1.586 (literature)
Linear Formula C4H3BrS
MDL Number MFCD00005417
Quantity 50 g
Synonym 2-Thienyl bromide
Molecular Weight (g/mol) 163.04
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.