Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020408 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0408 1 mL
11-101-8353 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0408 Supplier MilliporeSigma Supelco Supplier No. 968701MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 78-92-2
Vapor Pressure 12.5 mmHg (20°C)
Vapor Density 2.6 (vs. air)
Density 0.808 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH(OH)CH3
MDL Number MFCD00004569
Melting Point -115°C
Boiling Point 98°C
Synonym sec-Butyl alcohol
Quantity 1 mL
Formula Weight 74.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.