Learn More
Medchemexpress LLC 2-Butanone 2,4-dinitrophenylhydrazone-d3 | 259824-58-3 | 99.0% | C10H9D3N4O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY141854S1MG
Encompass_Preferred
2-Butanone 2,4-dinitrophenylhydrazone-d3 is the deuterium labeled 2-Butanone 2,4-dinitrophenylhydrazone.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has shown potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-53731