Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Butanone, 4-[4-(β-D-glucopyranosyloxy)phenyl]- | 38963-94-9 | 1 ML
SDP

Catalog No. 5000417756
Encompass_Preferred

Raspberry ketone glucoside is a melanogenesis inhibitor that activates the IL6/JAK1/STAT3 pathway, thereby inhibiting the transcriptional activity of MITFa and the downstream expression of TYR and TYRP1a genes. This compound demonstrates significant whitening activity in both B16F10 cells in vitro and in a zebrafish model in vivo.

  • Inhibits melanogenesis by activating the IL6/JAK1/STAT3 pathway
  • Inhibits transcriptional activity of MITFa
  • Reduces downstream expression of TYR and TYRP1a genes
  • Demonstrates whitening activity in B16F10 cells and zebrafish models
  • Appears as a white to off-white solid
  • Purity: 99.52%
  • Molecular formula: C16H22O7
  • Molecular weight: 326.34
  • Soluble in DMSO at 50 mg/mL with ultrasonication
  • Can be used as an analytical standard

Catalog No. 50-004-17756 Supplier Medchemexpress LLC Supplier No. HYN606910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raspberry ketone glucoside is a melanogenesis inhibitor that activates the IL6/JAK1/STAT3 pathway, thereby inhibiting the transcriptional activity of MITFa and the downstream expression of TYR and TYRP1a genes. This compound demonstrates significant whitening activity in both B16F10 cells in vitro and in a zebrafish model in vivo.

  • Inhibits melanogenesis by activating the IL6/JAK1/STAT3 pathway
  • Inhibits transcriptional activity of MITFa
  • Reduces downstream expression of TYR and TYRP1a genes
  • Demonstrates whitening activity in B16F10 cells and zebrafish models
  • Appears as a white to off-white solid
  • Purity: 99.52%
  • Molecular formula: C16H22O7
  • Molecular weight: 326.34
  • Soluble in DMSO at 50 mg/mL with ultrasonication
  • Can be used as an analytical standard

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.