Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020027 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0027 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0027 Supplier MilliporeSigma Supelco Supplier No. 024695ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Butanone also known as methyl ethyl ketone is a colorless fairly volatile liquid with a pleasant pungent odor. It is primarily used as a solvent, for making other chemicals, and for production of wax from petroleum. It is derived from refluxing methyl acetoacetate and dilute sulphuric acid. It finds application in manufacturing plastic, textile, paints, pharmaceuticals and cosmetics. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 78-93-3
Ignition Point 960°F
Vapor Pressure 71 mmHg (20°C)
Vapor Density 2.49 (vs. air)
Density 0.805 g/mL (at 25°C)
Flash Point -3°C
Linear Formula C2H5 COCH3
MDL Number MFCD00011648
Melting Point -87°C
Boiling Point 80°C
Synonym Ethyl methyl ketone; MEK; Methyl ethyl ketone
Type Analytical Standard
Quantity 5 mL
Formula Weight 72.11
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.