Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, 99+%, for electronic use (MOS), residue free

Catalog No. AC327910025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327910025 2.5 L Glass Bottle
AC327910010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327910025 Supplier Thermo Scientific Chemicals Supplier No. 327910025
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 78-93-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.8060g/mL
Quantity 2.5 L
Flash Point −7°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.0005% max.
Specific Gravity 0.806
Assay Percent Range 99+%
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Water 0.2% max. (Coulometric)
Beilstein 01, IV, 3243
Fieser 01,678
Merck Index 15, 6143
Boiling Point 80.0°C
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Electronic
Chemical Name or Material 2-Butanone, Residue Free
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.