Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, ACS Reagent, ≥99%, Solstice

Catalog No. 60046606 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-606 6 x 1 L Glass Bottle
60-046-601 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-606 Supplier Solstice Supplier No. 3604736X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -87°C
Density 0.805g/mL at 25°C
Quantity 6 x 1 L
Flash Point −3°C (26.6°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index n20/D 1.379
Boiling Point 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.