Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, for analysis ACS, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
78-93-3
C4H8O
72.107
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone,methyl ethyl ketone,butanone,ethyl methyl ketone,methylethyl ketone,meetco,methyl acetone,methylethylketone,3-butanone,ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(=O)C
78-93-3
C4H8O
72.107
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone,methyl ethyl ketone,butanone,ethyl methyl ketone,methylethyl ketone,meetco,methyl acetone,methylethylketone,3-butanone,ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(=O)C
Aluminum (Al) 0.5ppm max.
Linear Formula C2H5COCH3
Barium (Ba) 0.1ppm max.
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Cadmium (Cd) 0.05ppm max.
Calcium (Ca) 0.5ppm max.
Formula Weight 72.11
Physical Form Solution
Percent Purity ≥99.0% (GC)
Grade ACS Reagent
Chromium (Cr) 0.02ppm max.
Cobalt (Co) 0.02ppm max.
Copper (Cu) 0.02ppm max.
Density 0.8060g/mL
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.1ppm max.
Magnesium (Mg) 0.1ppm max.
Manganese (Mn) 0.02ppm max.
Nickel (Ni) 0.02ppm max.
Packaging Glass Bottle
Potassium (K) 0.1ppm max.
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.0025 % max.
Solubility Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Specific Gravity 0.806
Titratable Acid 0.0005 meq/g max.
Water 0.20% Max.
Zinc (Zn) 0.1ppm max.
Flash Point −7°C
Viscosity 0.42 mPa.s (15°C)
Boiling Point 80.0°C
Color Golden to Orange
Melting Point -87.0°C
Quantity 1L
Chemical Name or Material 2-Butanone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger