Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Solstice

Catalog No. 6000112 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-001-12 100 mL Glass Bottle
60-001-13 1 L Glass Bottle
60-001-14 2.5 L Glass Bottle
60-001-16 6 x 1 L Glass Bottle
60-001-15 4 x 2.5 L Glass Bottle
60-048-235 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-001-12 Supplier Solstice Supplier No. 34861100ML
Add to Cart
Add to Cart

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C (literature)
CAS Max % 100.0000%
Density 0.805 g/mL at 25 °C(lit.)
Quantity 100 mL
Flash Point −4°C
Packaging Glass Bottle
Linear Formula C2H5COCH3
UN Number UN1193
Beilstein 741880
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
Boiling Point 79°C to 81°C
Formula Weight 72.11
Percent Purity >99.7%
Grade HPLC
Chemical Name or Material 2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone
EINECSNumber 201-159-0
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.