Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone oxime, 99%

Catalog No. AC403330250
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403330250 25 g
AC403330010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403330250 Supplier Thermo Scientific Chemicals Supplier No. 403330250
  • $37.50
    $31.65 / Each of 1

    Save $5.85

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 96-29-7 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-29-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00013935
InChI Key WHIVNJATOVLWBW-SNAWJCMRSA-N
Synonym mek-oxime, butanone oxime, methyl ethyl ketone oxime, 2-butanone oxime, ethyl methyl ketoxime, usaf ek-906, ethyl-methylketonoxim, unii-51yge935u9, 2-butanone, oxime, methyl ethyl ketoxime
PubChem CID 5324276
IUPAC Name (NZ)-N-butan-2-ylidenehydroxylamine
SMILES CC\C(C)=N\O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9230g/mL
Boiling Point 72.0°C (25.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4410 to 1.4440
Linear Formula CH3CNOHC2H5
Quantity 25 g
Beilstein 01, IV, 3250
Specific Gravity 0.923
Solubility Information Solubility in water: 114g/L water (20°C).
Viscosity 15 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Butanone oxime, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Harmful in contact with skin.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-496-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.