Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600106 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-106 200 L Drum
18-600-098 100 mL Amber Glass Bottle
18-600-099 500 mL Amber Glass Bottle
18-600-101 1 L Amber Glass Bottle
18-600-100 6 x 500 mL Amber Glass Bottle
18-600-102 4 L Amber Glass Bottle
18-600-103 6 x 1 L Amber Glass Bottle
18-600-104 4 x 4 L Amber Glass Bottle
18-600-105 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-600-106 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1260200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1260, 78-93-3, C4H8O

Spectrum™ Chemical 2-Butanone, Reagent, ACS

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

CAS Max % 100%
Quantity 200 L
Packaging Drum
Residue after Evaporation 0.0025%
Titratable Acid 0.0005 meq/g
Water 0.2%
UN Number UN1193
Boiling Point 80 °C
Percent Purity 99%
Grade Reagent
Chemical Name or Material 2-Butanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.