Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Butanone suitable for HP
Catalog No. 502903924
Encompass
Physical properties of the SOLUTIONons of 1-n-butyl-3-methylimidazolium tetrafluoroborate (ionic liquid) in various organic solvents (acetonitrile dichloromethane 2-butanone and N N-dimethylformamide) has been studied. Viscosity and density of these mixtures were evaluated at 298.15K.
Catalog No. 50-290-3924 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348614X4L
Physical properties of the SOLUTIONons of 1-n-butyl-3-methylimidazolium tetrafluoroborate (ionic liquid) in various organic solvents (acetonitrile dichloromethane 2-butanone and N N-dimethylformamide) has been studied. Viscosity and density of these mixtures were evaluated at 298.15K.