Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, technical

Catalog No. AC444170050
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC444170050 5 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444170050 Supplier Thermo Scientific Chemicals Supplier No. 444170050
  • $115.00
    $91.65 / Each of 1

    Save $23.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 78-93-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
Density 0.8060g/mL
Quantity 5 L
Flash Point −7°C
Packaging Plastic Drum
Refractive Index 1.3780 to 1.3800
Specific Gravity 0.806
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Merck Index 15,6143
Boiling Point 80.0°C
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene,alcohol and ether,miscible with oils
Formula Weight 72.11
Grade Technical
Chemical Name or Material 2-Butanone
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 201-159-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.