Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Buten-1-amine, 3-fluoro-4-(phenylsulfonyl)-, hydrochloride (1:1), (2Z)- | 2409964-40-3 | C10H13ClFNO2S | 25 MG
SDP

Catalog No. 5000375992
Encompass_Preferred

PXS-4787 hydrochloride is a specific and effective pan-LOX (lysyl oxidase) inhibitor, abolishing lysyl oxidase activity. It inhibits LOX with IC50s ranging from 0.2 μM to 3.2 μM across various LOX isoforms, and also reduces deposition and crosslinking of collagen I secreted by human fibroblasts.

  • Purity of 99.71%
  • Solid, white to off-white appearance
  • Molecular weight of 265.73
  • Storage conditions: 4°C sealed for solid; -80°C for 6 months or -20°C for 1 month in solvent
  • Soluble in DMSO at 100 mg/mL
  • Inhibits LOX isoforms including Bovine LOX, rh LOXL1, rh LOXL2, rh LOXL3, and rh LOXL4
  • Reduces collagen I deposition and crosslinking

Catalog No. 50-003-75992 Supplier Medchemexpress LLC Supplier No. 25MGHY151498A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PXS-4787 hydrochloride is a specific and effective pan-LOX (lysyl oxidase) inhibitor, abolishing lysyl oxidase activity. It inhibits LOX with IC50s ranging from 0.2 μM to 3.2 μM across various LOX isoforms, and also reduces deposition and crosslinking of collagen I secreted by human fibroblasts.

  • Purity of 99.71%
  • Solid, white to off-white appearance
  • Molecular weight of 265.73
  • Storage conditions: 4°C sealed for solid; -80°C for 6 months or -20°C for 1 month in solvent
  • Soluble in DMSO at 100 mg/mL
  • Inhibits LOX isoforms including Bovine LOX, rh LOXL1, rh LOXL2, rh LOXL3, and rh LOXL4
  • Reduces collagen I deposition and crosslinking

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.