Learn More
Medchemexpress LLC 2-Butene-1,4-diamine, N1, N4-bis[3-(ethylamino)propyl]-, hydrochloride (1:4), (2Z)- | 206991-64-2 | 95.0% | 402.27 | 25 MG
SDP
Catalog No. 5000394816
Encompass_Preferred
PG-11047 (CGC-11047) is a polyamine analogue used for the research of breast cancer. It offers the following features and benefits:
  • Inhibits the growth of breast cell lines, with basal-like cell lines being inhibited at lower concentrations than luminal cell lines.
  • Causes a significant decrease in S phase fraction at doses that produce little apoptosis.
Catalog No. 50-003-94816 Supplier Medchemexpress LLC Supplier No. 25MGHY16394
PG-11047 (CGC-11047) is a polyamine analogue used for the research of breast cancer. It offers the following features and benefits:
  • Inhibits the growth of breast cell lines, with basal-like cell lines being inhibited at lower concentrations than luminal cell lines.
  • Causes a significant decrease in S phase fraction at doses that produce little apoptosis.