Learn More
Medchemexpress LLC 2-Butene-1,4-diamine, N1,N4-bis[3-(ethylamino)propyl]-, hydrochloride (1:4), (2Z)- | 206991-64-2 | 95.0% | 402.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1639450MG
Encompass_Preferred
PG-11047, also known as CGC-11047, is a polyamine analogue. It is utilized in the research of breast cancer. Studies have shown that it can inhibit the growth of breast cell lines, with basal-like cell lines being more sensitive. It also induces a significant decrease in the S phase fraction of cells at concentrations that produce minimal apoptosis. At higher concentrations, PG-11047 has been observed to induce apoptosis.
  • Polyamine analogue.
  • Used in breast cancer research.
  • Inhibits growth of breast cell lines.
  • More effective on basal-like breast cancer cell lines.
  • Decreases S phase fraction in cells.
  • Induces apoptosis at high concentrations.
Catalog No. 50-003-93748