Learn More
eMolecules​ 2-Butene-1,4-diol, cis/trans mixture | Combi-Blocks | 110-64-5 | | 88.106 | C4H8O2 | 95.000 | OCC=CCO | 25g | 303349993
Supplier:  eMolecules​ 303349993
Encompass_Preferred
2-Butene-1,4-diol, cis/trans mixture | Combi-Blocks | 110-64-5 | | 88.106 | C4H8O2 | 95.000 | OCC=CCO | 25g | 303349993
Catalog No. NC1421907