Learn More
Medchemexpress LLC 2-butenedioic-2,3-d2 acid, 1-methyl ester, (2E)- | 253122-44-0 | 98.2% | 132.11 | C5H4D2O4 | 5 MG
SDP
Catalog No. 5000247080
Encompass_Preferred
Monomethyl fumarate-d2 is the deuterium-labeled analogue of monomethyl fumarate, supplied as a solid research reagent for use in analytical, pharmacological, and metabolic studies. It serves as an internal standard and tracer in mass spectrometry and mechanistic experiments, and represents the active metabolite of dimethyl fumarate (DMF).
  • Deuterium labeled for isotope-tracing and mass spectrometry applications.
  • High purity suitable for analytical reference and research use.
  • Solid form for stable storage and handling.
  • Useful as a tracer in metabolic and pharmacokinetic studies.
  • CAS 253122-44-0 corresponds to the d2 isotopologue.
Catalog No. 50-002-47080 Supplier Medchemexpress LLC Supplier No. HY103252S25MG
Monomethyl fumarate-d2 is the deuterium-labeled analogue of monomethyl fumarate, supplied as a solid research reagent for use in analytical, pharmacological, and metabolic studies. It serves as an internal standard and tracer in mass spectrometry and mechanistic experiments, and represents the active metabolite of dimethyl fumarate (DMF).
  • Deuterium labeled for isotope-tracing and mass spectrometry applications.
  • High purity suitable for analytical reference and research use.
  • Solid form for stable storage and handling.
  • Useful as a tracer in metabolic and pharmacokinetic studies.
  • CAS 253122-44-0 corresponds to the d2 isotopologue.