Learn More
Medchemexpress LLC 2-Butenedioic acid (2E)-, 1,4-diethyl ester | 623-91-6 | 99.0% | 172.18 | 1 G
SDP
Catalog No. 5000366425
Encompass_Preferred
Diethyl fumarate is a decomposition product of Malathion 9, a pesticide, and also serves as a reactant in the Diels-Alder reaction. It is recognized as a skin irritant, capable of causing non-immunologic contact urticaria. Additionally, it has been shown to hybridize with curcumin, resulting in neuroprotective effects.
  • It is a decomposition product.
  • It acts as a reactant in Diels-Alder reactions.
  • It can cause non-immunologic contact urticaria on skin.
  • It can exert neuroprotective effects when hybridized with curcumin.
Catalog No. 50-003-66425 Supplier Medchemexpress LLC Supplier No. HYW0100561G
Diethyl fumarate is a decomposition product of Malathion 9, a pesticide, and also serves as a reactant in the Diels-Alder reaction. It is recognized as a skin irritant, capable of causing non-immunologic contact urticaria. Additionally, it has been shown to hybridize with curcumin, resulting in neuroprotective effects.
  • It is a decomposition product.
  • It acts as a reactant in Diels-Alder reactions.
  • It can cause non-immunologic contact urticaria on skin.
  • It can exert neuroprotective effects when hybridized with curcumin.