Learn More
Medchemexpress LLC 2-Butenedioic acid (2E)-, 1-ethyl ester | 2459-05-4 | MFCD00002699 | 99.6% | 144.13 g/mol | C6H8O4 | 1 ML
SDP
Catalog No. 5000300948
Encompass_Preferred
Monoethyl fumarate is the monoethyl ester of fumaric acid used as a research reagent. It acts as an activator of Nrf2 and is employed as a preservative and polymerization agent for macromolecular materials. The compound is available as a ready-to-use 10 mM solution in DMSO or as a bulk solid and is provided with analytical documentation confirming high purity.
Catalog No. 50-003-00948 Supplier Medchemexpress LLC Supplier No. HYW01969610MM/1ML
Monoethyl fumarate is the monoethyl ester of fumaric acid used as a research reagent. It acts as an activator of Nrf2 and is employed as a preservative and polymerization agent for macromolecular materials. The compound is available as a ready-to-use 10 mM solution in DMSO or as a bulk solid and is provided with analytical documentation confirming high purity.