Learn More
Medchemexpress LLC 2-Butenedioic acid (2E)-, 1-ethyl ester | 2459-05-4 | MFCD00002699 | 99.6% | 144.13 g/mol | C6H8O4 | 25 G
SDP
Catalog No. 5000303002
Encompass_Preferred
Monoethyl fumarate (2-Butenedioic acid (2E)-, 1-ethyl ester) is the monoethyl ester of fumaric acid supplied as a research reagent. It functions as an NRF2 activator and is used in biochemical and pharmacological studies to probe Keap1-Nrf2 signaling and inflammatory responses. Analytical documentation and high-purity material are provided for reproducible experiments.
  • High purity suitable for research applications.
  • Documented analytical data (COA, SDS, datasheet) available.
  • Useful as an NRF2 activator probe in Keap1-Nrf2 pathway studies.
  • Applicable to biochemical, pharmacological, and inflammation/immunology research.
  • Available in multiple sizes for flexible experimental scaling.
Catalog No. 50-003-03002 Supplier Medchemexpress LLC Supplier No. 25GHYW019696
Monoethyl fumarate (2-Butenedioic acid (2E)-, 1-ethyl ester) is the monoethyl ester of fumaric acid supplied as a research reagent. It functions as an NRF2 activator and is used in biochemical and pharmacological studies to probe Keap1-Nrf2 signaling and inflammatory responses. Analytical documentation and high-purity material are provided for reproducible experiments.
  • High purity suitable for research applications.
  • Documented analytical data (COA, SDS, datasheet) available.
  • Useful as an NRF2 activator probe in Keap1-Nrf2 pathway studies.
  • Applicable to biochemical, pharmacological, and inflammation/immunology research.
  • Available in multiple sizes for flexible experimental scaling.