Learn More
Medchemexpress LLC 2-butenedioic acid (2E)-, 1-ethyl ester | 2459-05-4 | MFCD00002699 | 99.6% | 144.13 g/mol | C6H8O4 | 50 G
SDP
Catalog No. 5000301488
Encompass_Preferred
Monoethyl fumarate is the monoethyl ester of fumaric acid supplied as a research reagent and reported Nrf2 activator. It is used in biochemical research and as a preservative and polymerization agent for macromolecular materials. The compound is available as a solid and as a 10 mM solution in DMSO for experimental convenience.
  • Activates Nrf2 signaling in research assays.
  • High reported purity (99.62%).
  • Molecular weight 144.13 g/mol and chemical formula C6H8O4.
  • Available in multiple pack sizes and solution form.
  • Useful as a preservative and polymerization agent for macromolecular materials.
  • For research use only; not for clinical use.
Catalog No. 50-003-01488 Supplier Medchemexpress LLC Supplier No. HYW01969650G
Monoethyl fumarate is the monoethyl ester of fumaric acid supplied as a research reagent and reported Nrf2 activator. It is used in biochemical research and as a preservative and polymerization agent for macromolecular materials. The compound is available as a solid and as a 10 mM solution in DMSO for experimental convenience.
  • Activates Nrf2 signaling in research assays.
  • High reported purity (99.62%).
  • Molecular weight 144.13 g/mol and chemical formula C6H8O4.
  • Available in multiple pack sizes and solution form.
  • Useful as a preservative and polymerization agent for macromolecular materials.
  • For research use only; not for clinical use.