Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Butenedioic acid (2E)- | 110-17-8 | MFCD00002700 | 116.07 g/mol | C4H4O4 | 25 G
SDP

Catalog No. 5000303081
Encompass_Preferred

Fumaric acid is an unsaturated dicarboxylic acid used in biochemical research as an endogenous metabolite and signaling modulator. Supplied as a solid for laboratory use, it is suitable for studies of metabolism and inflammatory signaling pathways such as NF-κB and p38 MAPK, and for preparing concentrated stock solutions for in vitro or in vivo experiments.

  • Molecular formula C4H4O4 and molecular weight 116.07 g/mol.
  • Soluble in DMSO and water; requires ultrasonic or warming for full dissolution.
  • Formulated for research use with storage guidance for solid and solvent solutions.
  • Suitable for metabolic, signaling, and pharmacological assays.

Catalog No. 50-003-03081 Supplier Medchemexpress LLC Supplier No. 25GHYW015883
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid is an unsaturated dicarboxylic acid used in biochemical research as an endogenous metabolite and signaling modulator. Supplied as a solid for laboratory use, it is suitable for studies of metabolism and inflammatory signaling pathways such as NF-κB and p38 MAPK, and for preparing concentrated stock solutions for in vitro or in vivo experiments.

  • Molecular formula C4H4O4 and molecular weight 116.07 g/mol.
  • Soluble in DMSO and water; requires ultrasonic or warming for full dissolution.
  • Formulated for research use with storage guidance for solid and solvent solutions.
  • Suitable for metabolic, signaling, and pharmacological assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.